Learn More
Medchemexpress LLC Carnosic acid (Standard) | 3650-09-7 | 332.43 | 5 MG
SDP
Catalog No. 5000388473
Encompass_Preferred
Carnosic acid (Standard) is an analytical standard intended for research and analytical applications. It has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity.
  • Intended for research and analytical applications
  • Inhibits oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Used as an analytical standard and reference standard for assays
  • Employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-88473 Supplier Medchemexpress LLC Supplier No. HYN0644R5MG
Add to Cart
Add to Cart
Carnosic acid (Standard) is an analytical standard intended for research and analytical applications. It has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity.
  • Intended for research and analytical applications
  • Inhibits oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Used as an analytical standard and reference standard for assays
  • Employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS