Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnosic Acid 97.0+%, TCI America™
SDP

Catalog No. C3477100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3477100MG 100 mg
C347720MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3477100MG Supplier TCI America Supplier No. C3477100MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3650-09-7
Molecular Formula C20H28O4
Molecular Weight (g/mol) 332.44
MDL Number MFCD02259459
InChI Key QRYRORQUOLYVBU-VBKZILBWSA-N
Synonym (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic Acid
PubChem CID 65126
ChEBI CHEBI:65585
IUPAC Name (4aR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid
SMILES CC(C)C1=C(C(=C2C(=C1)CCC3C2(CCCC3(C)C)C(=O)O)O)O

Specifications

Melting Point 190°C
Color Yellow
Quantity 100 mg
Formula Weight 332.44
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carnosic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.