Learn More
Medchemexpress LLC HY-N0643 5mg Medchemexpress, Carnosol CAS:5957-80-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06435MG
Encompass_Preferred
Medchemexpress, HY-N0643 5mg Carnosol CAS:5957-80-2 Carnosol is a potent Ribosomal S6 Kinase (RSK2) inhibitor that could be useful for treating gastric cancer, with an IC 50 of ~5.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3654