Learn More
Medchemexpress LLC HY-112431A 5mg Medchemexpress, Carnostatine (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112431A5MG
Encompass_Preferred
Medchemexpress, HY-112431A 5mg Carnostatine (HCl) CAS: Carnostatine hydrochloride (SAN9812 hydrochloride) is a potent and selective carnosinase 1 (CN1) inhibitor with a Ki of 11 nM for human recombinant CN1. Carnostatine hydrochloride can be used for the treatment of diabetic nephropathy (DN). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9092