Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Carnoy's 2000 Fixative
SDP

Catalog No. 21033368
Change view
Click to view available options
Grade:
Chemical Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
21-033-368 Chemical Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-368 Supplier StatLab Supplier No. FXCAR2PT
Add to Cart
Add to Cart

A low-hazard replacement for traditional Carnoys Fluid

  • Carnoys 2000TM fixes glycogen, Nissl substance, plasma cells, RNA, and DNA without chloroform, methanol, or mercuric chloride
  • It is compatible with routine processing alcohols and is intended for In Vitro Diagnostic use
  • Tissue fixed with Carnoys 2000 can be used for routine Special Stains and Immunohistochemical (IHC) stains

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.