Learn More
StatLab Carnoy's Fluid Fixative

Description
- It is particularly effective for chromosomal studies and tissue sample preparation
- The fixative is known for its ability to preserve delicate structures and is also used for DNA isolation in certain applications
- It is a fast fixative, making it suitable for rapid processing of paraffin sections and other tissue samples
Specifications
Specifications
| Chemical Name or Material | Carnoy's Fixative |
| CAS | 64-17-5, 64-19-7 |
| Grade | Laboratory |
| Color | Colorless |
| Quantity | 16 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.