Learn More
Carnoy's Solution (Fixative), Spectrum™ Chemical
SDP
Catalog No. 18601490 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Capacity:
500 mL
Catalog No. Capacity
18-601-490 500 mL
1 options
Catalog No. 18-601-490 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C475500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
C-475
Spectrum™ Chemical Carnoy's Solution (Fixative), a fixative composed of ethanol, chloroform and glacial acetic acid, can be used to enhance lymph node detection while dissecting cadavers and as a fixative for pap smear samples. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
Specifications
Capacity 500 mL
Packaging Type Amber Glass Bottle
Grade Chemical Solution