Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnoy's Solution (Fixative), Ricca Chemical
SDP

Catalog No. r18510004c Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R18510004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R18510004C Supplier Ricca Chemical Company Supplier No. R18510004C
Only null left
Add to Cart
Add to Cart

Specifications

Physical Form Liquid
Chemical Name or Material Carnoy's Solution
CAS 67-66-3, 64-17-5, 64-19-7, 67-63-0, 67-56-1
Packaging Amber Glass Bottle
Color Colorless
Quantity 4 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.