Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Carnoys 2000 Fixative
SDP

Catalog No. 22021405
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-405 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-405 Supplier StatLab Supplier No. FXCAR2GAL
Add to Cart
Edge
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Carnoys 2000 Fixative - a low-hazard replacement for traditional Carnoy's Fluid

  • Carnoy's 2000 fixes glycogen, Nissl substance, plasma cells, RNA and DNA without chloroform, methanol or mercuric chloride
  • Gallon size

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.