Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Animal Kingdom II Set
SDPSCI_ED

Catalog No. S22767ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S22767ND K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S22767ND Supplier Carolina™ Supplier No. POM1019
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Observe the diversity of the animal kingdom. The Animal Kingdom II Set represents 8 phyla, with 20 specimens embedded in a clear storage case.

  • Set represents 8 phyla
  • Collection of 20 specimens embedded in clear acrylic storage case
  • Includes color key

Specifications

Grade K to 12
Format Set
Product Type Animal Kingdom II Set
Includes Leech, cuttlefish, spider, centipede, crab, swordtail fish, frog, water snake, Lonchura (bird), bat, long-horned beetle, cricket, cockroach, wasp, lychee stink bug, cicada, mantis, butterfly, dragonfly, and stick insect.
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.