Learn More
Carolina™ Algae Food Source
SDPSCI_ED
Catalog No. S07459ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 quart
Catalog No. Quantity
S07459ND 1 quart
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07459ND Supplier Carolina™ Supplier No. 142337
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Freshwater Nannochloropsis grown in Alga-Gro™ Freshwater Medium.
Can be used for Daphnia but may also be used for other herbivorous crustaceans.
  • For filter feeding organisms such as Daphnia sp. and Moina sp.
  • 1 quart
Specifications
Quantity 1 quart