Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Animal Kingdom I Set
SDPSCI_ED

Catalog No. S22766ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S22766ND K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S22766ND Supplier Carolina™ Supplier No. POM1018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Observe the diversity of the animal kingdom. The Animal Kingdom I set includes a variety of species, as well as an identification key.

  • Collection of 12 representative specimens embedded in clear acrylic storage case
  • Includes: tortoise, leech, tiger barb fish, frog, paramecium, sea anemone, cuttlefish, scorpion, mouse, tapeworm, roundworm, and Zosterops (bird) and color key identifying specimens

Specifications

Grade K to 12
Format Set
Product Type Animal Kingdom I Set
Includes Tortoise, leech, tiger barb fish, frog, paramecium, sea anemone, cuttlefish, scorpion, mouse, tapeworm, roundworm and Zosterops (bird) specimens
Width (English) 13.6 in.
Height (English) 10.2 in.
Depth (English) 1.6 in.
Material Clear Acrylic
Languages English
For Use With (Application) Educational purposes
Subject Biology - Life Science
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.