missing translation for 'onlineSavingsMsg'
Learn More

Carolina™ Animal Kingdom Survey Set
SDPSCI_ED

Catalog No. S22782ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S22782ND K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S22782ND Supplier Carolina™ Supplier No. 261112
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Introduce students to forty-nine representative specimens. The Animal Kingdom Survey Set covers sponges, cnidarians, mollusks, annelids, echinoderms, myriapod, chelicerates, insects, crustaceans and vertebrates.

  • Forty-nine representative specimens covering the sponges, cnidarians, mollusks, annelids, echinoderms, myriapods, chelicerates, insects, crustaceans, and vertebrates are mounted
  • Mounted in twelve 6 x 5 in., glass-fronted Riker displays
  • Labeled

Specifications

Grade K to 12
Format Set
Product Type Animal Kingdom Survey Set
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.