Learn More
Carolina™ Basic Bacteriophage Culture Set A
SDPSCI_ED
Catalog No. S21829ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
S21829ND 1 Kit
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21829ND Supplier Carolina Biological Supply Supplier No. 121150
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes
Includes general information booklet
Recommended for introducing students to virulent bacterial viruses.
Knowledge of techniques in handling bacteriophage is essential in modern genetics and genetic engineering. Set contains 5mL each of high-titer phages T4 and T4r with one agar culture of E. coli B.
Specifications
Product Type Bacteriophage Culture
Quantity 1 Kit