Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Bioluminescent Bacterium Kit
SDPSCI_ED

Catalog No. s07430nd Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07430ND Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07430ND Supplier Carolina Biological Supply Supplier No. 154750

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • V. fischeri tube culture
  • 10 tubes of photobacterium agar
  • Metal inoculating loop
  • Instructions

Light up your students' understanding of bioluminescent organisms.

Students transfer Vibrio fischeri onto photobacterium agar, incubate the cultures in the dark at room temperature and observe bioluminescence the next day.

  • Easy to perform
  • Requires no experience in microbiology

Note: Store at room temperature; do not refrigerate or incubate. Use bacteria within one to two days of receipt for best results. Keep bacteria culture in envelope until use.

Specifications

For Use With (Application) Light up students understanding of bioluminescent organisms.
Storage Requirements 20°C to 22°C
Quantity Ea.
Includes V. fischeri tube culture, 10 tubes of photobacterium agar, metal inoculating loop, instructions
Sufficient For Class of 30 students
Product Type Bioluminescent Bacterium Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.