Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Butterflies in the Classroom Kit
SDPSCI_ED

Catalog No. S07707ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Includes:
Live Caterpillars
Prepaid Coupon for Caterpillars
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S07707ND Live Caterpillars
S07708ND Prepaid Coupon for Caterpillars
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07707ND Supplier Carolina Biological Supply Supplier No. 144014

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Painted Lady Butterfly Culture (33 larvae with food)
  • Painted Lady Butterfly Culture Medium (food)
  • Carolina Butterfly Sanctuary
  • 33 1 oz. cups with lids
  • Five wooden craft sticks
  • Five sorting brushes
  • 10 Kimwipes™
  • Foam flower
  • Cotton wick
  • Teacher's Manual and Reproducible Student Guide

Requires

Spray bottle; small bottle or vial; sugar; water; food coloring (optional); host plants (mallow, thistle, daisy, etc. optional)

These kits make keeping live butterflies simple, fun and affordable.

This kit allows students to work hands-on with living butterflies. From a culture cup that contains 33 larvae and food, each student (or pair of students) transfers a larva and food to its own culture vessel, creating an individual chamber for observation. The provided habitat is simple to set up, easy to clean, great for viewing and collapses for convenient storage.

Note: Prepaid coupons allow you to order your organisms in advance. We' ll send a coupon(s) that you later redeem by phone, fax, online Web form or mail. To ensure prompt arrival of material, coupons must be redeemed at least two weeks prior to requested delivery date.

Specifications

For Use With (Application) Allow students to work hands on with living butterflies.
Quantity Ea.
Includes Live Caterpillars
Sufficient For Class of 30 students
Product Type Butterflies in the Classroom Kit (with Live Caterpillars)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.