Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Coliphage T4r, Living, 5 mL
SDPSCI_ED

Catalog No. S21836ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21836ND 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21836ND Supplier Carolina Biological Supply Supplier No. 124335
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Rapid lysis; nonreverting plaque morphology mutant of T4.

Bacteriophage is individually tested for quality and titer. This coliphage is supplied as filtered (sterile) peptone broth suspension of high relative infective titer (109 to 1011 plaque-forming particles/mL) in screw-cap vial. Vial contains 5mL and proven titer is noted on label.

Specifications

Content And Storage Refrigerate coliphage until use; do not freeze.
Product Type Bacteriophage Culture
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.