Learn More
Carolina™ Duckweed
SDPSCI_ED
Catalog No. S07493ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
150/Container
Catalog No. Quantity
S07493ND 150/Container
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07493ND Supplier Carolina Biological Supply Supplier No. 161820
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Duckweed are small plants that often cover the water like a green carpet.
  • Lemna minor
  • Monocot
  • Unit contains 100 to 150 plants
  • With instructions
Specifications
Quantity 150/Container
Scientific Name Lemna minor
Includes 100 to 150 plants, instructions
Sufficient For Class of 30 students
Product Type Duckweed