Learn More
Description
- Lumbricus terrestris
- Large, mature specimens are suitable for laboratory study and dissection
- Medium-sized specimens are not suitable for dissection, but are good for making slides and for use as food for snakes and large salamanders
- With instructions
Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.
Specifications
Specifications
| For Use With (Application) | Laboratory study and dissection |
| Storage Requirements | 15°C |
| Quantity | 12/Container |
| Size | Large |
| Scientific Name | Lumbricus terrestris |
| Includes | Instructions |
| Sufficient For | Class of 15 students |
| Product Type | Earthworm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

