Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Earthworms
SDPSCI_ED

Catalog No. S07604ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
100/Container
12/Container
25/Container
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07604ND 100/Container
S07603ND 12/Container
S07271ND 25/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07604ND Supplier Carolina Biological Supply Supplier No. 141624

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Students can find these tube-shaped, segmented animals living in soil and feeding on live and dead organic matter.

  • Lumbricus terrestris
  • Large, mature specimens are suitable for laboratory study and dissection
  • Medium-sized specimens are not suitable for dissection, but are good for making slides and for use as food for snakes and large salamanders
  • With instructions

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

For Use With (Application) Laboratory study and dissection
Storage Requirements 15°C
Quantity 100/Container
Size Large
Scientific Name Lumbricus terrestris
Includes Instructions
Sufficient For Class of 60 students
Product Type Earthworm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.