Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Fern Allies Set
SDPSCI_ED

Catalog No. S07554ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
3/Set
Catalog No. Quantity
S07554ND 3/Set
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S07554ND

Supplier: Carolina Biological Supply 156950

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Selaginella
  • Lycopodium
  • Equisetum

Fern allies are a diverse group of seedless vascular plants that are not true ferns.

Specifications

For Use With (Application) Fern allies are a diverse group of seedless vascular plants that are not true ferns.
Storage Requirements 22°C to 25°C
Quantity 3/Set
Includes Selaginella, Lycopodium and Equisetum
Sufficient For Class of 30 students
Product Type Fern Allies Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.