Learn More
Carolina™ Carolina™ Formalin Grasshoppers
SDPSCI_ED
Catalog No. S72737 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Plain
Single
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type
S72737 Single
S72459 Plain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S72737 Supplier Carolina™ Supplier No. 225583
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carolina™ Formalin Grasshoppers are an excellent introductory specimen for dissecting and studying insect anatomy.
  • Louisiana Lubber Grasshoppers (Romalea microptera or Brachystola)
  • Perfect introductory dissection specimen
  • Large size is perfect for studying specialized external and internal anatomy
  • Preserved in formalin
  • Shipped 10 per jar
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid which minimizes student and educator exposure to formaldehyde
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Single
Quantity 10/Jar
Packaging Jar
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov