Learn More
Carolina™ Formalin Pig Eyes
SDPSCI_ED
Catalog No. S72455 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Bag
Catalog No. Quantity
S72455 10/Bag
1 options
Catalog No. S72455 Supplier Carolina™ Supplier No. 228551
Add to Cart
Add to Cart
Carolina™ Formalin Pig Eyes are for use in exploring the anatomy of mammalian eye. Carolina™ Formalin Pig Eyes are similar to human eye.
  • Easy dissection and examination of the mammalian eye as pig eyes are similar to human eye.
  • Smaller than cow eyes and have less muscle and fatty tissue attached
  • Preserved in formalin
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid which minimizes student and educator exposure to formaldehyde
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 10/Bag
Packaging Bulk Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov