Learn More
Carolina™ Formalin Rabbits
SDPSCI_ED
Catalog No. S72457 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1/Bag
Catalog No. Quantity
S72457 1/Bag
1 options
This item is not returnable. View return policy
Catalog No. S72457 Supplier Carolina™ Supplier No. 228220
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carolina™ Formalin Rabbits facilitates the study and dissection of mammalian anatomy
  • Can be used as a preserved cat alternative with minimal differences
  • Makes it easy to study musculature and internal anatomy
  • Mature specimens
  • Preserved in formalin
  • Shipped 1 per bag
  • Shipped in Carosafe,™ Carolina's proprietary shipping and holding fluid which minimizes student and educator exposure to formaldehyde
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Double
Length (English) 14+ in.
Quantity 1/Bag
Packaging Vac Pac
WARNING: Cancer - www.P65Warnings.ca.gov