Learn More
Carolina™ Formula 4-24™ Drosophila Medium
SDPSCI_ED
Catalog No. S07691ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Blue Formula 4-24 Instant Medium
Plain Formula 4-24 Instant Medium
Quantity:
4L
4 x 4L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
S07691ND Blue Formula 4-24 Instant Medium 4L
S07690ND Plain Formula 4-24 Instant Medium 4 x 4L
S07689ND Plain Formula 4-24 Instant Medium 4L
S07692ND Blue Formula 4-24 Instant Medium 4 x 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07691ND Supplier Carolina™ Supplier No. 173212
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes
  • Yeast
  • Measuring cups
  • Instructions
This medium allows teachers to set up a new culture in less than a minute.
  • Mold resistant
  • Needs no cooking or sterilizing
  • Just add water for a complete medium
  • Each liter of Formula 4-24™ will prepare 75 to 85 Drosophila culture vials
Specifications
Product Type Blue Formula 4-24 Instant Medium
For Use With (Application) Medium allows teachers to set up a new culture in less than a minute.
Includes Yeast, measuring cups and instructions
Quantity 4L