Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formula 4-24™ Drosophila Medium
SDPSCI_ED

Catalog No. s07689nd Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Blue Formula 4-24 Instant Medium
Plain Formula 4-24 Instant Medium
Quantity:
4L
4 x 4L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
S07689ND Plain Formula 4-24 Instant Medium 4L
S07691ND Blue Formula 4-24 Instant Medium 4L
S07690ND Plain Formula 4-24 Instant Medium 4 x 4L
S07692ND Blue Formula 4-24 Instant Medium 4 x 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S07689ND Supplier Carolina™ Supplier No. 173202
Only null left
Add to Cart
Add to Cart

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Yeast
  • Measuring cups
  • Instructions

This medium allows teachers to set up a new culture in less than a minute.

  • Mold resistant
  • Needs no cooking or sterilizing
  • Just add water for a complete medium
  • Each liter of Formula 4-24™ will prepare 75 to 85 Drosophila culture vials

Specifications

Autoclavable Not Autoclavable
Product Type Plain Formula 4-24 Instant Medium
Color Natural
For Use With (Application) Medium allows teachers to set up a new culture in less than a minute.
Includes Yeast, measuring cups and instructions
Quantity 4L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.