Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Gram Stain Comparison Set
SDPSCI_ED

Catalog No. S07420ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
5/Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07420ND 5/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07420ND Supplier Carolina Biological Supply Supplier No. 154615

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Bacillus cereus (Gram-positive rod)
  • Branhamella catarrhalis (Gram-negative coccus)
  • Micrococcus luteus (Gram-positive coccus)
  • Rhodospirillum rubrum (Gram-negative spiral)
  • Serratia marcescens (Gram-negative rod)

A selection of five bacterial tube cultures representing Gram-negative and Gram-positive bacteria. This characteristic of bacteria is determined by the make-up of the cell wall, tested by using the Gram-stain method, which is used as the first step in identifying a bacterium.

Specifications

Packaging Tube
Storage Requirements 20°C to 22°C
Quantity 5/Set
Includes Bacillus cereus (Gram-positive rod), Branhamella catarrhalis (Gram-negative coccus), Micrococcus luteus (Gram-positive coccus), Rhodospirillum rubrum (Gram-negative spiral), Serratia marcescens (Gram-negative rod)
Sufficient For Class of 30 students
Product Type Gram Stain Comparison Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.