Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Amoeba, Paramecium and Euglena Review Set
SDPSCI_ED

Catalog No. S07444ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
3/Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07444ND 3/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07444ND Supplier Carolina Biological Supply Supplier No. 131085

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • A separate culture of Amoeba, Paramecium and Euglena
  • A pad of 30 Bioreview™ sheets for each culture
  • Instructions for culturing each form
  • Materials for a class of 30

The protozoan body consists of a single cell, but it is a mistake to think of these organisms as simple.

Specifications

Storage Requirements 20°C to 22°C
Quantity 3/Set
Includes Separate culture of Amoeba, Paramecium and Euglena, pad of 30 Bioreview™ sheets for each culture, instructions for culturing each form
Sufficient For Class of 30 students
Product Type Amoeba, Paramecium, Euglena Review Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.