Learn More
Carolina™ Bacillus coagulans, Living, Tube
SDPSCI_ED
Catalog No. S21888ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Tube
Catalog No. Quantity
S21888ND 1 Tube
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21888ND Supplier Carolina Biological Supply Supplier No. 154885
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
This culture is Gram+, motile rods, acid tolerant, and causes food spoilage.
It is cultured on tryptic soy or nutrient agar at 37 degrees C in a tube. Cultures are guaranteed as to species and purity.
  • Each culture contains enough material for a class of 30 students.
Specifications
Storage Requirements 37°C
Quantity 1 Tube
Media Type Tryptic Soy Agar or Nutrient Agar
Product Type Bacteria Culture
Gram Stain Result Gram (+)