Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Bacillus sphaericus, Living, Plate
SDPSCI_ED

Catalog No. S21889ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21889ND 1 Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21889ND Supplier Carolina Biological Supply Supplier No. 154908

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Cultures are guaranteed as to species and purity.

Cultures are labeled by genus and species name and media on which they are cultured. All Gram staining based on 24-hr cultures.

  • Enough material for a class of 30 students

Specifications

Storage Requirements 22°C
Quantity 1 Tube
Scientific Name Bacillus sphaericus
Sufficient For Class of 30 students
Media Type Nutrient Agar
Product Type Bacteria
Temperature (Metric) Range 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.