Learn More
Carolina™ Bacillus thuringiensis, Living, Tube
SDPSCI_ED
Catalog No. S21891ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Tube
Catalog No. Quantity
S21891ND 1 Tube
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21891ND Supplier Carolina Biological Supply Supplier No. 154926
 Restricted Item
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
This culture is Gram+, motile rods, and crystals within cell body pathogenic to some insects (Lepidoptera).
It is cultured on nutrient agar at 30 degrees C in a tube. Cultures are guaranteed as to species and purity.
  • Each culture contains enough material for a class of 30 students.
Specifications
Storage Requirements 30°C
Quantity 1 Tube
Media Type Nutrient Agar
Product Type Bacteria Culture
Gram Stain Result Gram (+)