Learn More
Description
- Wild-type strain
- Extensively used for genetic studies
- Aseptically maintained on Nematode Growth Agar with E. coli as the feeder host
Specifications
Specifications
| For Use With (Application) | Wild type strain, extensively used for genetic studies. |
| Packaging | Plate |
| Storage Requirements | 20°C |
| Quantity | Ea. |
| Scientific Name | Caenorhabditis elegans N2 |
| Sufficient For | Class of 30 students |
| Product Type | Bacteria Culture |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

