Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Chromobacterium violaceum, MicroKwik Culture™, PATHOGEN, Vial
SDPSCI_ED

Catalog No. S21818ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21818ND 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21818ND Supplier Carolina Biological Supply Supplier No. 154931A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

Note: Each MicroKwik Culture™ comes with 5mL of rehydration medium.

This Biosafety Level 2 (pathogen) culture is freeze-dried cells plus medium.

Every culture is guaranteed for species and purity. Bacteria cultures are labeled by genus and species name, media on which they are cultivated, and temperature for optimal growth. All Gram staining is based on 24-hour cultures. Special packing and transportation costs are extra for animal and human pathogens.

  • Each culture contains enough material for a class of 30 students
  • Gram - , motile rods, with violet pigment
  • Cultured on nutrient agar at 30°C

Specifications

Quantity 1 Vial
Media Type Nutrient Agar
Product Type Bacteria Culture
Gram Stain Result Gram (-)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.