Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Daphnia Food
SDPSCI_ED

Catalog No. S07593ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
4 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07593ND 4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07593ND Supplier Carolina Biological Supply Supplier No. 142316

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Use to culture D. pulex and D. magna in both small and large volumes.

  • For small volumes, use 1/16 tsp. (0.2g) per gallon
  • For large volumes, use 1/2 tsp. (1g) per 5 gallons

Specifications

For Use With (Application) For culturing D. pulex and D. magna in both small and large volumes.
Storage Requirements 24°C to 31°C
Quantity 4 oz.
Sufficient For Class of 30 students
Product Type Daphnia Food
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.