Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Daphnia magna
SDPSCI_ED

Catalog No. S07268CND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Daphnia Magna
Daphnia Magna Coupon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07268CND Daphnia Magna Coupon
S07268ND Daphnia Magna
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S07268CND Supplier Carolina Biological Supply Supplier No. TF142330LC
Add to Cart
Add to Cart

Cultures of D. magna contain adults, juveniles, and neonates.

  • For a class of 30 students
  • A large species of Daphnia
  • Commonly used in aquatic effluent toxicity testing
  • An obligate algal feeder
  • Cultured using powdered Spirulina as a food source

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

For Use With (Application) Commonly used in aquatic effluent toxicity testing, obligate algal feeder.
Storage Requirements 21°C
Quantity Ea.
Scientific Name Daphnia magna
Includes Adults, juveniles, neonates
Sufficient For Class of 30 students
Product Type Daphnia Magna Coupon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.