Learn More
Carolina™ Drosophila Chromosome 1 Mutants
SDPSCI_ED
Catalog No. S07682ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Drosophila, White
Catalog No. Product Type
S07682ND Drosophila, White
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07682ND Supplier Carolina Biological Supply Supplier No. 172220
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Students can easily study the sex-linked, recessive and dominant mutations.
Drosophila yields many flies from just one vial of media. Chromosome 1 Mutants (sex-linked) are easy-to-score strains which are excellent for beginning genetics. These are recommended also for investigating sex-linked inheritance.
  • Short generation times
  • Sequenced genome
  • Affordable
  • Minimal storage space
  • Easy for beginners to score
Specifications
For Use With (Application) Students can easily study the sex-linked, recessive and dominant mutations, recommended also for investigating sex-linked inheritance.
Storage Requirements 20°C to 25°C
Quantity Ea.
Product Type Drosophila, White