Learn More
Description
This vigorous, healthy plant needs good light and should be anchored in soil or sand.
- Cultivated form
- Keeps water conditioned and oxygenated
- Can survive and grow as floating fragments
- Recommended for photosynthesis studies
- With instructions
Specifications
Specifications
| For Use With (Application) | This vigorous, healthy plant needs good light and should be anchored in soil or sand, keeps water conditioned and oxygenated, can survive and grow as floating fragments, recommended for photosynthesis studies. |
| Quantity | 25/Container |
| Scientific Name | Elodea canadensis |
| Includes | Instructions |
| Sufficient For | Class of 25 students |
| Product Type | Elodea canadensis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

