Learn More
Carolina™ Elodea canadensis
SDPSCI_ED
Catalog No. p-4655629 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
12/Container
25/Container
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07494ND 12/Container
S07495ND 25/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07494ND Supplier Carolina Biological Supply Supplier No. 162111
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Elodea canadensis is similar to Elodea densa but much smaller.
This vigorous, healthy plant needs good light and should be anchored in soil or sand.
  • Cultivated form
  • Keeps water conditioned and oxygenated
  • Can survive and grow as floating fragments
  • Recommended for photosynthesis studies
  • With instructions
Specifications
For Use With (Application) Keeps water conditioned and oxygenated, can survive and grow as floating fragments, recommended for photosynthesis studies.
Quantity 12/Container
Scientific Name Elodea canadensis
Includes Instructions
Sufficient For Class of 15 students
Product Type Elodea canadensis