Learn More
Carolina™ Escherichia coli B, Living, Bacteriophage Host
SDPSCI_ED
Catalog No. S21831ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
S21831ND 1 Kit
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21831ND Supplier Carolina Biological Supply Supplier No. 124300
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
An optimum host organism for all T coliphages.
These bacteria are cultured on tryptone agar slants in screw-cap tubes.
Specifications
Product Type Bacteriophage Host
Quantity 1 Kit