Learn More
Carolina™ Escherichia coli K12, Living, Bacteriophage Host
SDPSCI_ED
Catalog No. S21839ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Tube
Catalog No. Quantity
S21839ND 1 Tube
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21839ND Supplier Carolina Biological Supply Supplier No. 124500
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Lysogenic for coliphage lambda.
Supports reproduction of wild-type phage T4 resulting in Iysis, but T4rll mutants are unable to form plaques on this bacterial strain. These bacteria are cultured on tryptone agar slants in screw-cap tubes.
Specifications
Product Type Bacteriophage Host
Quantity 1 Tube