Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Hydra
SDPSCI_ED

Catalog No. S07265ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Hydra
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07265ND Hydra
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07265ND Supplier Carolina Biological Supply Supplier No. 132800

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

An underwater wonder!

Generally, the brown hydra species shipped is Hydra littoralis. Specimens are freshly collected to ensure maximum natural size and optimum physiological condition.

  • Easily maintained in a freshwater aquarium
  • For a class of 30 students
  • Needs no filtration or aeration system
  • Reproduces rapidly when fed daily

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

Packaging Jar
Storage Requirements 21°C
Quantity Ea.
Sufficient For Class of 30 students
Product Type Hydra
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.