Learn More
Description
Includes
- A separate culture of Hydra, Planaria and Daphnia
- A pad of 30 Bioreview™ Sheets for each
- Instructions for culturing each form
Specifications
Specifications
| For Use With (Application) | To observe the similarities and differences between some of the most popular invertebrates. |
| Storage Requirements | 21°C |
| Quantity | 3/Set |
| Includes | A separate culture of Hydra, Planaria and Daphnia, a pad of 30 Bioreview™ sheets for each, instructions for culturing each form |
| Sufficient For | Class of 30 students |
| Product Type | Hydra, Planaria And Daphnia Review Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

