Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Ludwigia
SDPSCI_ED

Catalog No. S07502ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
12/Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07502ND 12/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07502ND Supplier Carolina Biological Supply Supplier No. 162141

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

A very attractive plant to use in aquariums and bog terrariums.

  • False loosestrife
  • Leaves vary from green to red
  • With instructions
  • Container of 12

Specifications

For Use With (Application) Use in aquariums and bog terrariums.
Quantity 12/Container
Includes Instructions
Sufficient For Class of 30 students
Product Type Ludwigia
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.