Learn More
Description
A freshwater annelid that is excellent for studying regeneration, locomotion, circulation, and behavior.
- Lumbriculus variegatus
- For a class of 30 students
- One to two inch worms are more “user friendly” than either Planaria or Daphnia
- Easy to handle, manipulate, and culture in the lab
- With complete care instructions
Specifications
Specifications
| For Use With (Application) | Excellent for studying regeneration, locomotion, circulation and behavior. |
| Quantity | Ea. |
| Scientific Name | Lumbriculus variegatus |
| Includes | Instructions |
| Sufficient For | Class of 30 students |
| Product Type | Lumbriculus variegatus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

