Learn More
Description
Includes
- 100 larvae
- Care instructions
Larvae also demonstrate the life cycle of an insect with complete metamorphosis.
- Sarcophaga bullata
- Mature lab-grown larvae
- Pupate after a few days and flies emerge in 1 to 2 weeks
- Can use for forensics activities or as live food for reptiles and amphibians
Specifications
Specifications
| For Use With (Application) | Can use for forensics activities or as live food for reptiles and amphibians. |
| Storage Requirements | 27°C |
| Quantity | 100/Container |
| Scientific Name | Sarcophaga bullata |
| Includes | 100 larvae, care instructions |
| Sufficient For | Class of 30 students |
| Product Type | Sarcophaga Larvae |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

