Learn More
Carolina™ Sordaria Demonstration Cross Plate
SDPSCI_ED
Catalog No. S07440ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
Ea.
Catalog No. Quantity
S07440ND Ea.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07440ND Supplier Carolina Biological Supply Supplier No. 155846
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
A cross of Sordaria fimicola wild type and the mutant tan strain for demonstrating genetic crossing-over.
  • Easy to perform
  • Requires some basic training in microbiology
  • One plate
Specifications
For Use With (Application) For demonstrating genetic crossing-over, for use with AP™ biology laboratory 3 or as a separate refresher demonstration from the exercises found in Sordaria genetics kits.
Quantity Ea.
Product Type Sordaria Demonstration Cross Plate