Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Bacterial Cultures (A-C)
SDPSCI_ED

Catalog No. S07402ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Packaging:
Tube
Vial
Scientific Name:
Alcaligenes faecalis
Bacillus cereus
Bacillus megaterium
Bacillus stearothermophilus
Bacillus subtilis
Citrobacter freundii
Clostridium sporogenes
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Scientific Name
S07402ND Vial Bacillus megaterium
S07185ND Tube Bacillus stearothermophilus
S07406ND Vial Clostridium sporogenes
S07182ND Tube Bacillus cereus
S07416ND Tube Citrobacter freundii
S07423ND Vial Alcaligenes faecalis
S07184ND Tube Bacillus subtilis
S07425ND Vial Bacillus cereus
S07424ND Vial Bacillus subtilis
S07183ND Tube Bacillus megaterium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07402ND Supplier Carolina Biological Supply Supplier No. 154900A

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.

Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.

  • Labeled by genus and species name and media on which they are cultured
  • All Gram staining based on 24 hr. cultures
  • Pure and high-quality
  • For every level of study
  • Easy-to-use media

‡ This is a human or animal pathogen. It cannot be shipped to high schools, preparatory schools, or individuals, in accordance with guidelines of the U.S. Department of Health and Human Services.

Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.

Specifications

For Use With (Application) Ideal for food and chemical industries because of their highly diverse metabolic activities.
Packaging Vial
Storage Requirements 20°C to 22°C
Quantity Ea.
Scientific Name Bacillus megaterium
Includes Carolina™ techniques for studying bacteria and fungi manual
Sufficient For Class of 30 students
Media Type Nutrient Agar
Product Type Bacteria MicroKwik Culture™
Temperature (Metric) Range 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.