Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Bacterial Cultures (D-M)
SDPSCI_ED

Catalog No. S07418ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Bacteria Culture
Bacteria Culture Plate
Bacteria Culture Tube
Bacteria MicroKwik Culture
Scientific Name:
E.coli
Enterobacter aerogenes
Enterococcus faecalis
Klebsiella pneumoniae
Micrococcus luteus
Micrococcus roseus
Mycobacterium phlei
Mycobacterium smegmatis
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Scientific Name
S07418ND Bacteria Culture Plate E.coli
S07417ND Bacteria Culture Tube E.coli
S07210ND Bacteria Culture Micrococcus roseus
S07408ND Bacteria MicroKwik Culture Enterococcus faecalis
S07192ND Bacteria Culture Micrococcus luteus
S07413ND Bacteria MicroKwik Culture Klebsiella pneumoniae
S07180ND Bacteria Culture Enterobacter aerogenes
S07193ND Bacteria MicroKwik Culture Mycobacterium smegmatis
S07407ND Bacteria MicroKwik Culture Enterobacter aerogenes
S07194ND Bacteria MicroKwik Culture Mycobacterium phlei
S07403ND Bacteria MicroKwik Culture E.coli
S07189ND Bacteria Culture E.coli
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07418ND Supplier Carolina Biological Supply Supplier No. 155067

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.

Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.

  • Labeled by genus and species name and media on which they are cultured
  • All Gram staining based on 24 hr. cultures
  • Pure and high-quality
  • For every level of study
  • Easy-to-use media

‡ This is a human or animal pathogen. It cannot be shipped to high schools, preparatory schools, or individuals, in accordance with guidelines of the U.S. Department of Health and Human Services.

Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.

Specifications

For Use With (Application) Ideal for food and chemical industries because of their highly diverse metabolic activities.
Packaging Plate
Storage Requirements 20°C to 22°C
Quantity Ea.
Scientific Name E.coli
Includes Carolina™ techniques for studying bacteria and fungi manual
Sufficient For Class of 30 students
Media Type Nutrient Agar
Product Type Bacteria Culture Plate
Temperature (Metric) Range 37°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.