Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Bacterial Cultures (N-Z)
SDPSCI_ED

Catalog No. S07429ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Packaging:
Tube
Vial
Scientific Name:
Proteus mirabilis
Proteus vulgaris
Pseudomonas aeruginosa
Pseudomonas fluorescens
Salmonella typhimurium
Serratia marcescens
Serratia marcescens D1
Shigella flexneri
Staphylococcus aureus (Coagulase positive)
Staphylococcus epidermidis
Streptococcus pneumoniae
Streptococcus pyogenes
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Scientific Name
S07429ND Tube Staphylococcus epidermidis
S07410ND Vial Pseudomonas fluorescens
S07203ND Vial Staphylococcus aureus (Coagulase positive)
S07196ND Vial Proteus vulgaris
S07195ND Vial Pseudomonas aeruginosa
S07202ND Tube Serratia marcescens D1
S07414ND Vial Salmonella typhimurium
S07197ND Tube Pseudomonas fluorescens
S07409ND Vial Proteus mirabilis
S07411ND Vial Shigella flexneri
S07405ND Vial Staphylococcus epidermidis
S07201ND Tube Serratia marcescens
S07426ND Vial Serratia marcescens
S07401ND Vial Streptococcus pneumoniae
S07400ND Vial Streptococcus pyogenes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07429ND Supplier Carolina Biological Supply Supplier No. 155556

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.

Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.

  • Labeled by genus and species name and media on which they are cultured
  • All Gram staining based on 24 hr. cultures
  • Pure and high-quality
  • For every level of study
  • Easy-to-use media

‡ This is a human or animal pathogen. It cannot be shipped to high schools, preparatory schools, or individuals, in accordance with guidelines of the U.S. Department of Health and Human Services.

Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.

Specifications

For Use With (Application) Ideal for food and chemical industries because of their highly diverse metabolic activities.
Packaging Tube
Storage Requirements 20°C to 22°C
Quantity Ea.
Scientific Name Staphylococcus epidermidis
Includes Carolina™ techniques for studying bacteria and fungi manual
Sufficient For Class of 30 students
Media Type Nutrient Agar
Product Type Bacteria Culture
Temperature (Metric) Range 37°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.