Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Ciliate Cultures
SDPSCI_ED

Catalog No. S07224ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Paramecium
Paramecium caudatum
Stentor coeruleus
Vorticella
Packaging:
Jar
Scientific Name:
Paramecium
Paramecium caudatum
Stentor coeruleus
Vorticella
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Scientific Name
S07224ND Vorticella Jar Vorticella
S07219ND Paramecium Jar Paramecium
S07220ND Paramecium caudatum Jar Paramecium caudatum
S07223ND Stentor coeruleus Jar Stentor coeruleus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07224ND Supplier Carolina Biological Supply Supplier No. 131660

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Most ciliates are heterotrophs, feeding on smaller organisms, such as bacteria and algae.

Note: To ensure you order enough living materials for the number of students in your class, order 1 culture for up to 30 students, 2 cultures for 31 to 60 students, 3 cultures for 61 to 90 students, etc.

Specifications

Packaging Jar
Storage Requirements 20°C to 22°C
Quantity Ea.
Scientific Name Vorticella
Includes Dropping pipet, appropriate medium
Sufficient For Class of 30 students
Media Type Not recommended for culturing
Product Type Vorticella
Temperature (Metric) Range 22°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.